siRNA Details
Gene Symbol: Ifi44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107729.1 | NP_001101199.1 | 8 | 1379 | ||
XM_006233495.2 | XP_006233557.1 | 8 | 1407 | |||
XM_006233496.2 | XP_006233558.1 | 8 | 1337 | |||
GenBank mRNA | BC088420.1 | - | 9 | 1403 |
