siRNA Details
Gene Symbol: Ifi44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107729.1 | NP_001101199.1 | 5, 6 | 1044 | ||
XM_006233495.2 | XP_006233557.1 | 5, 6 | 1072 | |||
XM_006233496.2 | XP_006233558.1 | 5, 6 | 1002 | |||
XM_006233497.2 | XP_006233559.1 | 5, 6 | 1072 | |||
XM_006233498.2 | XP_006233560.1 | 5, 6 | 1002 | |||
GenBank mRNA | BC088420.1 | - | 6, 7 | 1068 | ||
BC158758.1 | - | 5, 6 | 936 | 7099624 |

More Information
Other Species Targeted

