siRNA Details
Gene Symbol: Rbm43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037649.1 | NP_001032738.1 | 5 | 769 | ||
XM_006234167.2 | XP_006234229.1 | 5 | 833 | |||
XM_006234168.2 | XP_006234230.1 | 5 | 1631 | |||
XM_008761743.1 | XP_008759965.1 | 5 | 1635 | |||
XM_008761744.1 | XP_008759966.1 | 5 | 837 | |||
GenBank mRNA | BC107648.1 | - | 5 | 769 | 7930358 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
