siRNA Details
Gene Symbol: Dppa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_236761.3 | XP_236761.1 | 2 | 874 |
Gene Symbol: RGD1564306 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_229957.4 | XP_229957.2 | 1 | 276 |
