siRNA Details
Gene Symbol: Psmd14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025689.1 | NP_001020860.1 | 8 | 771 | ||
GenBank mRNA | BC097427.1 | - | 8 | 771 | ||
BC158645.1 | - | 8 | 719 | 8363877 | ||
FQ228827.1 | - | 8 | 747 |

More Information
Other Species Targeted

Gene Ontology Categories:
