siRNA Details
Gene Symbol: Psmd14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025689.1 | NP_001020860.1 | 8, 9 | 859 | ||
GenBank mRNA | BC097427.1 | - | 8, 9 | 859 | ||
BC158645.1 | - | 8, 9 | 807 | 8363877 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
