siRNA Details
Gene Symbol: Cobll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234301.2 | XP_006234363.1 | 11 | 3462 | ||
XM_006234304.2 | XP_006234366.1 | 10 | 3356 | |||
XM_008761934.1 | XP_008760156.1 | 11 | 2268 | |||
XM_008761935.1 | XP_008760157.1 | 11 | 2253 | |||
XM_008761936.1 | XP_008760158.1 | 10 | 2229 | |||
XM_008761937.1 | XP_008760159.1 | 11 | 2268 | |||
XM_008761938.1 | XP_008760160.1 | 12 | 3503 | |||
XM_008761939.1 | XP_008760161.1 | 10 | 2163 | |||
XM_008761940.1 | XP_008760162.1 | 10 | 2148 | |||
XM_008761941.1 | XP_008760163.1 | 11 | 3464 | |||
XM_008761942.1 | XP_008760164.1 | 11 | 3856 | |||
XM_008761943.1 | XP_008760165.1 | 10 | 3358 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
