siRNA Details
Gene Symbol: Cobll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234301.2 | XP_006234363.1 | 13 | 3880 | ||
XM_006234304.2 | XP_006234366.1 | 12 | 3774 | |||
XM_008761934.1 | XP_008760156.1 | 13 | 2686 | |||
XM_008761935.1 | XP_008760157.1 | 13 | 2671 | |||
XM_008761936.1 | XP_008760158.1 | 12 | 2647 | |||
XM_008761937.1 | XP_008760159.1 | 13 | 2686 | |||
XM_008761938.1 | XP_008760160.1 | 14 | 3921 | |||
XM_008761939.1 | XP_008760161.1 | 12 | 2581 | |||
XM_008761940.1 | XP_008760162.1 | 12 | 2566 | |||
XM_008761941.1 | XP_008760163.1 | 13 | 3882 | |||
XM_008761942.1 | XP_008760164.1 | 13 | 4274 | |||
XM_008761943.1 | XP_008760165.1 | 12 | 3776 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
