siRNA Details
Gene Symbol: Clp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009599.1 | NP_001009599.1 | 2 | 148 | ||
XM_006234461.2 | XP_006234523.1 | 2 | 683 | |||
XM_006234462.1 | XP_006234524.1 | 2 | 231 | |||
GenBank mRNA | BC087130.1 | - | 2 | 148 | 7112583 | |
FQ231160.1 | - | 2 | 175 |
