siRNA Details
Gene Symbol: Slc43a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107742.1 | NP_001101212.1 | 3 | 490 | ||
GenBank mRNA | BC161926.1 | - | 3 | 451 | 8373798 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc43a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107742.1 | NP_001101212.1 | 3 | 490 | ||
GenBank mRNA | BC161926.1 | - | 3 | 451 | 8373798 |