siRNA Details
Gene Symbol: Rtn4rl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181380.2 | NP_852045.1 | 2 | 213 | ||
XM_006234465.2 | XP_006234527.1 | 2 | 525 | |||
XM_006234466.2 | XP_006234528.1 | 2 | 1136 | |||
XM_008761989.1 | XP_008760211.1 | 2 | 708 | |||
XM_008761990.1 | XP_008760212.1 | 2 | 313 | |||
GenBank mRNA | AF532860.1 | - | 2 | 213 | ||
AX751537.1 | - | 2 | 195 |
