siRNA Details
Gene Symbol: Ckap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234604.2 | XP_006234666.1 | 21 | 2622 | ||
XM_008762040.1 | XP_008760262.1 | 21 | 2703 | |||
XM_008762041.1 | XP_008760263.1 | 21 | 3276 | |||
XM_008762042.1 | XP_008760264.1 | 17 | 2141 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
