siRNA Details
Gene Symbol: Lrrc4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107753.1 | NP_001101223.1 | 3268 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_178725, BC094588, AK032467, AK032088, AK048322, and others.
Gene Ontology Categories:
