siRNA Details
Gene Symbol: Aven | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234707.2 | XP_006234769.1 | 2 | 387 | ||
XM_008762093.1 | XP_008760315.1 | 2 | 387 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
