siRNA Details
Gene Symbol: Meis2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107758.1 | NP_001101228.1 | 5 | 714 | ||
XM_006234742.2 | XP_006234804.1 | 5 | 912 | |||
XM_006234743.2 | XP_006234805.1 | 6 | 924 | |||
XM_006234744.2 | XP_006234806.1 | 6 | 958 | |||
XM_006234745.2 | XP_006234807.1 | 6 | 890 | |||
XM_006234746.2 | XP_006234808.1 | 5 | 912 | |||
XM_006234747.2 | XP_006234809.1 | 5 | 701 | |||
XM_006234748.2 | XP_006234810.1 | 5 | 1035 | |||
XM_006234749.2 | XP_006234811.1 | 5 | 701 | |||
XM_006234750.2 | XP_006234812.1 | 5 | 912 | |||
XM_006234751.2 | XP_006234813.1 | 5 | 912 | |||
XM_006234752.2 | XP_006234814.1 | 5 | 701 | |||
XM_006234753.2 | XP_006234815.1 | 5 | 1035 | |||
XM_006234754.2 | XP_006234816.1 | 5 | 701 | |||
XM_006234755.2 | XP_006234817.1 | 5 | 1035 | |||
XM_008762094.1 | XP_008760316.1 | 2 | 206 |

More Information
Other Species Targeted

Gene Ontology Categories:
