siRNA Details
Gene Symbol: Disp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107759.2 | NP_001101229.2 | 8 | 1986 | ||
XM_006234756.2 | XP_006234818.1 | 8 | 2000 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
