siRNA Details
Gene Symbol: Ttbk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107766.1 | NP_001101236.1 | 8 | 861 | ||
XM_006234833.2 | XP_006234895.1 | 10 | 1294 | |||
XM_006234834.2 | XP_006234896.1 | 10 | 937 | |||
XM_006234835.2 | XP_006234897.1 | 9 | 1220 | |||
XM_006234836.2 | XP_006234898.1 | 8 | 1072 | |||
XM_006234837.2 | XP_006234899.1 | 10 | 940 |

More Information
Other Species Targeted

Gene Ontology Categories:
