siRNA Details
Gene Symbol: Adal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014047.1 | NP_001014069.1 | 1, 2 | 286 | ||
XM_008762166.1 | XP_008760388.1 | 6, 7 | 764 | |||
XM_008762167.1 | XP_008760389.1 | 4, 5 | 294 | |||
XM_008762168.1 | XP_008760390.1 | 2, 3 | 195 | |||
XM_008762169.1 | XP_008760391.1 | 5, 6 | 500 | |||
GenBank mRNA | BC089959.1 | - | 1, 2 | 286 | 7313149 |
