siRNA Details
Gene Symbol: Wdr76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234874.2 | XP_006234936.1 | 11 | 1983 | ||
XM_006234875.2 | XP_006234937.1 | 10 | 1890 | |||
XM_006234876.2 | XP_006234938.1 | 10 | 1428 | |||
XM_006234877.1 | XP_006234939.1 | 10 | 1421 | |||
XM_008762207.1 | XP_008760429.1 | 7 | 1100 | |||
XM_008762208.1 | XP_008760430.1 | 8 | 1022 | |||
XM_008762209.1 | XP_008760431.1 | 7 | 849 | |||
XM_008762210.1 | XP_008760432.1 | 7 | 787 | |||
XM_230512.7 | XP_230512.4 | 10 | 1459 |
