siRNA Details
Gene Symbol: Ctdspl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014048.1 | NP_001014070.1 | 3 | 322 | ||
GenBank mRNA | BC083672.1 | - | 3 | 322 | 7135706 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ctdspl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014048.1 | NP_001014070.1 | 3 | 322 | ||
GenBank mRNA | BC083672.1 | - | 3 | 322 | 7135706 |