siRNA Details
Gene Symbol: Ctdspl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014048.1 | NP_001014070.1 | 12 | 1400 | ||
GenBank mRNA | BC083672.1 | - | 12 | 1400 | 7135706 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_212450, AK049463, AK035067, BC066219, AK053513, and others.
Gene Ontology Categories:
