siRNA Details
Gene Symbol: Eif3j | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077670.1 | NP_001071138.1 | 5 | 453 | ||
GenBank mRNA | BC127509.1 | - | 5 | 453 | 7454886 | |
FQ226401.1 | - | 5 | 402 |

More Information
Other Species Targeted

Gene Ontology Categories:
