siRNA Details
Gene Symbol: Usp50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749567.3 | XP_003749615.1 | 5 | 968 | ||
XM_006234940.2 | XP_006235002.1 | 5 | 1042 | |||
XM_006234941.2 | XP_006235003.1 | 5 | 1167 | |||
XM_006234942.2 | XP_006235004.1 | 5 | 1093 | |||
XM_006234943.2 | XP_006235005.1 | 5 | 683 | |||
XM_008762231.1 | XP_008760453.1 | 5 | 765 |
