siRNA Details
Gene Symbol: Mtfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100977.1 | NP_001094447.1 | 3 | 282 | ||
XM_006232178.2 | XP_006232240.1 | 4 | 439 | |||
XM_006232179.2 | XP_006232241.1 | 4 | 418 | |||
XM_006232180.2 | XP_006232242.1 | 3 | 1228 | |||
GenBank mRNA | EF221637.1 | - | 3 | 282 | ||
FQ217237.1 | - | 4 | 390 |
