siRNA Details
Gene Symbol: Zc3h8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012090.1 | NP_001012090.1 | 7 | 870 | ||
GenBank mRNA | BC079122.1 | - | 7 | 870 | 7126707 |

More Information
Other Species Targeted

Gene Ontology Categories:
