siRNA Details
Gene Symbol: Spef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039024.2 | NP_001034113.2 | 2 | 271 | ||
XM_006235033.2 | XP_006235095.1 | 2 | 2625 | |||
GenBank mRNA | BC099127.1 | - | 2 | 271 | 7457521 |

More Information
Other Species Targeted

