siRNA Details
Gene Symbol: Mavs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005556.1 | NP_001005556.1 | 2, 3 | 405 | ||
XM_006235034.2 | XP_006235096.1 | 2, 3 | 273 | |||
XM_006235035.2 | XP_006235097.1 | 2, 3 | 987 | |||
XM_006235036.2 | XP_006235098.1 | 3, 4 | 306 | |||
XM_008762179.1 | XP_008760401.1 | 2, 3 | 1480 | |||
XM_008762180.1 | XP_008760402.1 | 2, 3 | 293 | |||
GenBank mRNA | BC081869.1 | - | 2, 3 | 405 | 7106026 |
