siRNA Details
Gene Symbol: Mavs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005556.1 | NP_001005556.1 | 6 | 1247 | ||
XM_006235034.2 | XP_006235096.1 | 6 | 1115 | |||
XM_006235035.2 | XP_006235097.1 | 6 | 1829 | |||
XM_006235036.2 | XP_006235098.1 | 7 | 1148 | |||
XM_006235037.2 | XP_006235099.1 | 5 | 1151 | |||
XM_008762179.1 | XP_008760401.1 | 6 | 2322 | |||
XM_008762180.1 | XP_008760402.1 | 6 | 1135 | |||
GenBank mRNA | BC081869.1 | - | 6 | 1247 | 7106026 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
