siRNA Details
Gene Symbol: Rassf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037096.1 | NP_001032173.1 | 3 | 289 | ||
XM_006235066.2 | XP_006235128.1 | 3 | 972 | |||
GenBank mRNA | BC107656.1 | - | 3 | 289 | 7935449 |
