siRNA Details
Gene Symbol: Gzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107788.1 | NP_001101258.1 | 4 | 1808 | ||
XM_006235153.2 | XP_006235215.1 | 4 | 1675 | |||
XM_006235154.2 | XP_006235216.1 | 4 | 2787 | |||
XM_008762274.1 | XP_008760496.1 | 5 | 1966 |
