siRNA Details
Gene Symbol: Slc52a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037198.1 | NP_001032275.1 | 1 | 423 | ||
XM_006235238.2 | XP_006235300.1 | 4 | 1094 | |||
XM_008762327.1 | XP_008760549.1 | 4 | 1474 | |||
XM_008762328.1 | XP_008760550.1 | 1 | 1285 | |||
XM_008762329.1 | XP_008760551.1 | 1 | 1286 | |||
GenBank mRNA | BC099097.1 | - | 1 | 423 | 7459579 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
