siRNA Details
Gene Symbol: Tpx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107790.1 | NP_001101260.1 | 11 | 1701 | ||
XM_006235270.2 | XP_006235332.1 | 12 | 1604 | |||
XM_006235272.2 | XP_006235334.1 | 12 | 1556 | |||
XM_006235273.2 | XP_006235335.1 | 11 | 1486 | |||
XM_006235274.2 | XP_006235336.1 | 12 | 1604 |

More Information
Other Species Targeted

Gene Ontology Categories:
