siRNA Details
Gene Symbol: Pofut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002278.1 | NP_001002278.1 | 3 | 420 | ||
XM_006235290.2 | XP_006235352.2 | 3 | 564 | |||
GenBank mRNA | AJ781499.1 | - | 3 | 420 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
