siRNA Details
Gene Symbol: Phf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107795.1 | NP_001101265.1 | 5, 6 | 617 | ||
XM_006235404.2 | XP_006235466.1 | 6, 7 | 693 | |||
XM_008762342.1 | XP_008760564.1 | 6, 7 | 704 | |||
GenBank mRNA | FQ228046.1 | - | 2, 3 | 149 | ||
FQ232700.1 | - | 1, 2 | 200 | |||
FQ232914.1 | - | 2, 3 | 149 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_172674, AK045309, AK033017, AK038573, BC060121.
Gene Ontology Categories:
