siRNA Details
Gene Symbol: Chd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107797.1 | NP_001101267.1 | 17 | 2596 | ||
XM_006235482.2 | XP_006235544.1 | 16 | 2837 | |||
XM_006235483.2 | XP_006235545.1 | 16 | 3253 | |||
XM_006235484.2 | XP_006235546.1 | 16 | 2837 | |||
XM_008762345.1 | XP_008760567.1 | 17 | 2616 | |||
XM_008762346.1 | XP_008760568.1 | 16 | 2561 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_173368, BC150806, AK080892, AK018129, AK040495, and others.
Gene Ontology Categories:
