siRNA Details
Gene Symbol: Slc35c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107803.1 | NP_001101273.1 | 7 | 957 | ||
XM_006235588.2 | XP_006235650.1 | 8 | 1831 | |||
XM_006235590.2 | XP_006235652.1 | 8 | 1706 | |||
XM_008762504.1 | XP_008760726.1 | 8 | 1716 | |||
XM_008762505.1 | XP_008760727.1 | 7 | 2635 | |||
GenBank mRNA | BC087085.1 | - | 7 | 1681 | ||
BC166837.1 | - | 8 | 1000 | 9027110 | ||
FQ233415.1 | - | 8 | 1058 |
