siRNA Details
Gene Symbol: Ssna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233613.1 | XP_006233675.1 | 2 | 316 | ||
GenBank mRNA | BC166994.1 | - | 2 | 232 | 7304458 | |
CB576781.1 | - | 2 | 269 | |||
FN805842.1 | - | 2 | 211 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
