siRNA Details
Gene Symbol: Agpat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107821.2 | NP_001101291.1 | 2 | 304 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Agpat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107821.2 | NP_001101291.1 | 2 | 304 |