siRNA Details
Gene Symbol: Urm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137562.2 | NP_001131034.2 | 5 | 736 | ||
GenBank mRNA | BC169094.1 | - | 5 | 726 | 7933748 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
