siRNA Details
Gene Symbol: Crat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004085.2 | NP_001004085.1 | 6 | 897 | ||
XM_006233823.1 | XP_006233885.1 | 7 | 1417 | |||
XM_008761663.1 | XP_008759885.1 | 7 | 900 | |||
XM_008761664.1 | XP_008759886.1 | 6 | 766 | |||
GenBank mRNA | AJ620886.1 | - | 6 | 675 | ||
BC083616.1 | - | 6 | 897 | 7133847 | ||
FQ230824.1 | - | 7 | 1012 |
