siRNA Details
Gene Symbol: Fibcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107829.1 | NP_001101299.1 | 4 | 914 |
Gene Symbol: NEWGENE_1309097 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761689.1 | XP_008759911.1 | 4 | 890 |
