siRNA Details
Gene Symbol: Ptges2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107832.1 | NP_001101302.1 | 2 | 818 | ||
GenBank mRNA | FQ231959.1 | - | 2 | 311 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_133783, AK050616, AK182234, BC004846, AK002798.
Gene Ontology Categories:
