siRNA Details
Gene Symbol: Fbxw2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107835.2 | NP_001101305.1 | 3 | 661 | ||
XM_006233976.1 | XP_006234038.1 | 3 | 795 | |||
XM_006233977.2 | XP_006234039.1 | 4 | 711 | |||
XM_006233980.2 | XP_006234042.1 | 4 | 619 | |||
XM_008761750.1 | XP_008759972.1 | 4 | 2270 | |||
XM_008761751.1 | XP_008759973.1 | 5 | 876 | |||
XM_008761752.1 | XP_008759974.1 | 3 | 597 | |||
GenBank mRNA | BC166990.1 | - | 3 | 661 | 7110795 |

More Information
Other Species Targeted

