siRNA Details
Gene Symbol: Lhx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107837.1 | NP_001101307.1 | 5 | 700 | ||
XM_006234066.2 | XP_006234128.1 | 5 | 901 | |||
XM_008761753.1 | XP_008759975.1 | 5 | 962 |

More Information
Other Species Targeted

Gene Ontology Categories:
