siRNA Details
Gene Symbol: Rc3h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107839.1 | NP_001101309.1 | 18 | 3097 | ||
XM_008761754.1 | XP_008759976.1 | 18 | 3198 | |||
XM_008761755.1 | XP_008759977.1 | 17 | 2892 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
