siRNA Details
Gene Symbol: Rc3h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107839.1 | NP_001101309.1 | 10, 11 | 1514 | ||
XM_008761754.1 | XP_008759976.1 | 10, 11 | 1615 |

More Information
Other Species Targeted

Gene Ontology Categories:
