siRNA Details
Gene Symbol: Dennd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191747.1 | NP_001178676.1 | 21 | 1898 | ||
XM_008761756.1 | XP_008759978.1 | 22 | 2754 | |||
XM_008761757.1 | XP_008759979.1 | 22 | 2691 |
