siRNA Details
Gene Symbol: Golga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107842.1 | NP_001101312.1 | 5 | 573 | ||
XM_006234096.2 | XP_006234158.1 | 5 | 627 | |||
XM_006234097.2 | XP_006234159.1 | 5 | 583 | |||
XM_006234099.2 | XP_006234161.1 | 5 | 627 | |||
XM_008761758.1 | XP_008759980.1 | 5 | 629 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029793, AK008972, BC024637, AK044987, AK002954, and others.
Gene Ontology Categories:
