siRNA Details
Gene Symbol: Abcf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109666.1 | NP_001103136.1 | 11 | 1539 | ||
XM_006235929.2 | XP_006235991.1 | 12 | 1509 | |||
XM_006235930.2 | XP_006235992.1 | 12 | 1671 | |||
XM_006235931.2 | XP_006235993.1 | 12 | 1506 | |||
XM_006235932.2 | XP_006235994.1 | 12 | 1500 | |||
XM_006235933.2 | XP_006235995.1 | 12 | 1497 | |||
XM_006235934.2 | XP_006235996.1 | 11 | 1419 | |||
GenBank mRNA | BC129119.1 | - | 11 | 1406 | 7115823 |

More Information
Other Species Targeted

Gene Ontology Categories:
