siRNA Details
Gene Symbol: Abcf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109666.1 | NP_001103136.1 | 6, 7 | 1102 | ||
XM_006235929.2 | XP_006235991.1 | 7, 8 | 1072 | |||
XM_006235930.2 | XP_006235992.1 | 7, 8 | 1234 | |||
XM_006235931.2 | XP_006235993.1 | 7, 8 | 1069 | |||
XM_006235932.2 | XP_006235994.1 | 7, 8 | 1063 | |||
XM_006235933.2 | XP_006235995.1 | 7, 8 | 1060 | |||
XM_006235934.2 | XP_006235996.1 | 7, 8 | 1072 | |||
GenBank mRNA | BC129119.1 | - | 6, 7 | 969 | 7115823 |
